This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05003978_s1
See other ELAVL3 GE Assays ›
Gene Symbol:
ELAVL3
Gene Name:
ELAV like neuron-specific RNA binding protein 3
Gene Aliases:
HUC, HUCL, PLE21
Chromosome Location:
Chr.19: 11451328 - 11480994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05003978_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001420.3 NP_001411.2 7 2467 100
NM_032281.2 NP_115657.2 7 2446 100
GenBank mRNA BC011875.2 - 1 214 3161837 100
BC080533.1 - 1 212 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC011875.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ELAVL3

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
cell differentiation
nucleotide binding
AU-rich element binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000588853.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000588853.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000588853.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000592218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000592218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 ENST00000592218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000359227.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000438662.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000588853.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000588853.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000588853.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000592218.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000592218.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 ENST00000592218.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_001420.3 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq
Affymetrix Human Clariom D Assay TC1900011870.hg.1 NM_032281.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0