This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05004432_m1
See other CEACAM19 GE Assays ›
Gene Symbol:
CEACAM19
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 19
Gene Aliases:
CEACM19, CEAL1
Chromosome Location:
Chr.19: 44671452 - 44684355 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05004432_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127893.1 NP_001121365.1 4 - 5 1139 63
NM_020219.3 NP_064604.2 4 - 5 1139 63
GenBank mRNA AK027157.1 - 4 - 5 856 63
AK128234.1 - 1 - 2 1350 63
AK291588.1 - 4 - 5 840 63
AY358394.1 - 4 - 5 869 63
BC083499.1 - 4 - 5 1139 30390224 63
KJ851726.1 - 4 - 5 659 63
KJ851727.1 - 4 - 5 659 63
KJ851728.1 - 4 - 5 659 63
KJ851729.1 - 4 - 5 659 63
KJ851730.1 - 4 - 5 659 63
KJ851731.1 - 4 - 5 659 63
KJ851742.1 - 3 - 4 508 63
KJ851743.1 - 3 - 4 508 63
KJ851744.1 - 3 - 4 508 63
KJ851745.1 - 3 - 4 508 63
KJ851746.1 - 3 - 4 508 63
KJ851747.1 - 3 - 4 508 63
KJ851748.1 - 3 - 4 508 63
KJ851749.1 - 3 - 4 508 63
KJ851760.1 - 5 - 6 793 63
KJ851761.1 - 5 - 6 793 63
KJ851762.1 - 5 - 6 793 63
KJ851763.1 - 5 - 6 793 63
KJ851764.1 - 5 - 6 793 63
KJ851765.1 - 5 - 6 793 63
KJ851766.1 - 5 - 6 793 63
KJ851767.1 - 5 - 6 793 63
KJ851778.1 - 4 - 5 642 63
KJ851779.1 - 4 - 5 642 63
KJ851780.1 - 4 - 5 642 63
KJ851781.1 - 4 - 5 642 63
KJ851782.1 - 4 - 5 642 63
KJ851783.1 - 4 - 5 642 63
KJ851784.1 - 4 - 5 642 63
KJ851785.1 - 4 - 5 642 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CEACAM19

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007652.hg.1 ENST00000618435.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007652.hg.1 ENST00000618435.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5lprg:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0