This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006245_m1
See other MTRF1 GE Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
MRF1, MTTRF1, RF1
Chromosome Location:
Chr.13: 41216369 - 41311794 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05006245_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTRF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011535323.2 XP_011533625.1 2 - 3 160 104
XM_017020865.1 XP_016876354.1 1 - 2 1171 104
XM_017020874.1 XP_016876363.1 2 - 3 160 104
XM_017020879.1 XP_016876368.1 1 - 2 1171 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011535323.2, XM_017020865.1, XM_017020874.1, XM_017020879.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

translation release factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of translational termination
mitochondrial translational termination
translation release factor activity
translation release factor activity, codon specific
ribosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0