This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006253_m1
See other ESD GE Assays ›
Gene Symbol:
ESD
Gene Name:
esterase D
Gene Aliases:
FGH
Chromosome Location:
Chr.13: 46771256 - 46797681 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05006253_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001984.1 NP_001975.1 8 - 9 782 101
XM_005266278.2 XP_005266335.1 8 - 9 846 101
XM_011534954.1 XP_011533256.1 8 - 9 734 101
XM_017020414.1 XP_016875903.1 8 - 9 2056 101
GenBank mRNA AF112219.1 - 8 - 9 782 101
AK093643.1 - 2 - 3 171 101
AK098540.1 - 8 - 9 711 101
BC001169.1 - 8 - 9 697 3528593 101
BT007059.1 - 6 - 7 599 101
M13450.1 - 7 - 8 644 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ESD

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
formaldehyde catabolic process
glutathione derivative biosynthetic process
protein binding
hydrolase activity, acting on ester bonds
S-formylglutathione hydrolase activity
methylumbelliferyl-acetate deacetylase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0