This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006343_m1
See other SACS GE Assays ›
Gene Symbol:
SACS
Gene Name:
sacsin molecular chaperone
Gene Aliases:
ARSACS, DNAJC29, PPP1R138, SPAX6
Chromosome Location:
Chr.13: 23328823 - 23433728 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05006343_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SACS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278055.1 NP_001264984.1 7 - 8 2269 95
NM_014363.5 NP_055178.3 9 - 10 2800 95
XM_017020539.1 XP_016876028.1 8 - 9 2408 95
GenBank mRNA AK125458.1 - 7 - 8 2269 95
BX640926.1 - 5 - 6 1883 95
CR749427.1 - 2 - 3 1037 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SACS

Gene Ontology Categories:

Function(s) Process(es)

protein folding
negative regulation of inclusion body assembly
Hsp70 protein binding
chaperone binding
proteasome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382292.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000382298.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000402364.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000423156.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000455470.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 ENST00000476776.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_001278055.1 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq
Affymetrix Human Clariom D Assay TC1300008329.hg.1 NM_014363.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0