This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006346_m1
See other TNFRSF19 GE Assays ›
Gene Symbol:
TNFRSF19
Gene Name:
TNF receptor superfamily member 19
Gene Aliases:
TAJ, TAJ-alpha, TRADE, TROY
Chromosome Location:
Chr.13: 23570248 - 23676105 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05006346_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011535146.1 XP_011533448.1 2 - 3 187 103
XM_017020651.1 XP_016876140.1 2 - 3 690 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011535146.1, XM_017020651.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

hair follicle development
apoptotic process
JNK cascade
tissue development
tumor necrosis factor-mediated signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JNK cascade
receptor activity
tumor necrosis factor-activated receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0