This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006449_m1
See other FREM2 GE Assays ›
Gene Symbol:
FREM2
Gene Name:
FRAS1 related extracellular matrix protein 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 38687036 - 38887131 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05006449_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FREM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207361.5 NP_997244.4 13 - 14 7527 100
GenBank mRNA AK127571.1 - 10 - 11 1775 100
BX538304.1 - 9 - 10 1491 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FREM2

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
cell communication
cell adhesion
heart development
inner ear development
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0