This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05007587_s1
See other GPR33 GE Assays ›
Gene Symbol:
GPR33
Gene Name:
G protein-coupled receptor 33 (gene/pseudogene)
Gene Aliases:
-
Chromosome Location:
Chr.14: 31482944 - 31487945 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05007587_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197184.2 NP_001184113.2 2 871 75
NR_036675.2 2 871 75
GenBank mRNA GQ981321.1 - 2 871 75
GQ981322.1 - 2 871 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline