This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05008788_s1
See other GTF2A1 GE Assays ›
Gene Symbol:
GTF2A1
Gene Name:
general transcription factor IIA subunit 1
Gene Aliases:
TF2A1, TFIIA, TFIIA-42, TFIIAL
Chromosome Location:
Chr.14: 81175452 - 81221231 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05008788_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278940.1 NP_001265869.1 10 2790 85
NM_015859.3 NP_056943.1 9 2723 85
NM_201595.2 NP_963889.1 9 2343 85
GenBank mRNA BC041410.2 - 1 122 85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC037828.1 - 1 939 Chr. 14 - 81176140 - 81180000 [-] on Build GRCh38 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2A1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
DNA binding
transcription coactivator activity
protein binding
transcription factor binding
TBP-class protein binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0