This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05010526_mH
See other LOC100128108 GE Assays ›
Gene Symbol:
LOC100128108
Gene Name:
putative ubiquitin-conjugating enzyme E2Q2-like protein
Gene Aliases:
-
Chromosome Location:
Chr.15: 101736747 - 101746870 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05010526_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100128108
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321551.1 NP_001308480.1 2 - 3 246 91
NM_001321552.1 NP_001308481.1 2 - 3 260 91
NM_001321553.1 NP_001308482.1 1 - 2 329 91
XM_017021852.1 XP_016877341.1 2 - 3 587 91
GenBank mRNA AI417995.1 - 1 - 2 91
HY056262.1 - 2 - 3 260 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000559812.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000559812.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010673.hg.1 ENST00000558592.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010673.hg.1 ENST00000558592.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010673.hg.1 ENST00000558592.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010673.hg.1 ENST00000560292.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010673.hg.1 ENST00000560292.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010673.hg.1 ENST00000560292.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000622345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000622345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010797.hg.1 ENST00000622345.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010673.hg.1 ENST00000558592.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010673.hg.1 ENST00000558592.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010673.hg.1 ENST00000558592.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010673.hg.1 ENST00000560292.2 Ensembl
Affymetrix Human Clariom D Assay TC1500010673.hg.1 ENST00000560292.2 Ensembl
Affymetrix Human Clariom D Assay TC1500010673.hg.1 ENST00000560292.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000557887.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000558345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000559812.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000559812.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560306.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 ENST00000560917.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038273.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500008153.hg.1 NR_038274.1 Refseq
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000560059.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000622345.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000622345.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010797.hg.1 ENST00000622345.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0