This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05010559_m1
See other GLCE GE Assays ›
Gene Symbol:
GLCE
Gene Name:
glucuronic acid epimerase
Gene Aliases:
HSEPI
Chromosome Location:
Chr.15: 69160555 - 69272209 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05010559_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLCE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324091.1 NP_001311020.1 2 - 3 249 88
NM_001324093.1 NP_001311022.1 3 - 4 376 88
NM_001324094.1 NP_001311023.1 3 - 4 1376 88
NM_015554.2 NP_056369.1 2 - 3 249 88
XM_005254298.3 XP_005254355.1 3 - 4 381 88
GenBank mRNA AY635582.1 - 2 - 3 212 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY635582.1, NM_001324091.1, NM_001324093.1, NM_001324094.1, NM_015554.2, XM_005254298.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLCE

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
racemase and epimerase activity, acting on carbohydrates and derivatives
heparosan-N-sulfate-glucuronate 5-epimerase activity
UDP-glucuronate 5'-epimerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 ENST00000584174.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000261858.6 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559420.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559500.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559500.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559500.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000559798.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 ENST00000584174.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq
Affymetrix Human Clariom D Assay TC1500007691.hg.1 NM_015554.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0