This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05011271_s1
See other ISLR GE Assays ›
Gene Symbol:
ISLR
Gene Name:
immunoglobulin superfamily containing leucine rich repeat
Gene Aliases:
HsT17563, Meflin
Chromosome Location:
Chr.15: 74173746 - 74176871 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05011271_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ISLR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005545.3 NP_005536.1 2 999 134
NM_201526.1 NP_958934.1 2 815 134
GenBank mRNA AB003184.1 - 2 815 134
AB024536.1 - 2 999 134
AK000817.1 - 2 999 134
AK074668.1 - 2 1407 134
AK223267.1 - 2 831 134
AK297730.1 - 2 579 134
AY358871.1 - 2 1075 134
BC022478.1 - 2 804 4794738 134
BC111013.1 - 2 909 5763157 134

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ISLR

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

platelet degranulation
cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000559510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000559510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000560862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000560862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 ENST00000560862.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000249842.7 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000395118.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000559510.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000559510.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000560862.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000560862.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 ENST00000560862.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_005545.3 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_201526.1 Refseq
Affymetrix Human Clariom D Assay TC1500007839.hg.1 NM_201526.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline