This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05014864_s1
See other NRM GE Assays ›
Gene Symbol:
NRM
Gene Name:
nurim (nuclear envelope membrane protein)
Gene Aliases:
NRM29
Chromosome Location:
Chr.6: 30688047 - 30691420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05014864_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NRM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270707.1 NP_001257636.1 4 1398 84
NM_001270708.1 NP_001257637.1 3 1225 84
NM_001270709.1 NP_001257638.1 3 1203 84
NM_001270710.1 NP_001257639.1 2 1006 84
NM_007243.2 NP_009174.1 5 1577 84
NR_073065.1 4 1402 84
NR_073066.1 3 1183 84
NR_073072.1 3 1046 84
XM_017010224.1 XP_016865713.1 4 1579 84
XM_017010225.1 XP_016865714.1 5 1493 84
GenBank mRNA AF143676.1 - 4 1223 84
AI521967.1 - 1 84
AK075509.1 - 5 1579 84
AK298152.1 - 3 1040 84
AY358411.1 - 4 1244 84
BC004509.2 - 2 849 84
BC021260.1 - 4 1215 84
BC039865.1 - 4 1243 6043663 84
JQ798186.1 - 3 1066 84
JQ798187.1 - 2 869 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NRM

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0