This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05015040_s1
See other BIRC5 GE Assays ›
Gene Symbol:
BIRC5
Gene Name:
baculoviral IAP repeat containing 5
Gene Aliases:
API4, EPR-1
Chromosome Location:
Chr.17: 78214196 - 78225635 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05015040_s1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BIRC5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012270.1 NP_001012270.1 3 1351 72
NM_001012271.1 NP_001012271.1 5 1538 72
NM_001168.2 NP_001159.2 4 1469 72
GenBank mRNA AF077350.1 - 4 1372 72
AK223428.1 - 4 1415 72
AL519719.3 - 1 44 72
BC000784.2 - 3 1270 72
BC008718.2 - 4 1381 2961114 72
BC034148.1 - 4 1386 4656567 72
BC065497.1 - 4 1361 5394399 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BIRC5

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
protein phosphorylation
apoptotic process
sister chromatid cohesion
positive regulation of cell proliferation
regulation of signal transduction
protein ubiquitination
protein sumoylation
protein complex localization
regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
cell division
mitotic spindle assembly
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
ubiquitin-protein transferase activity
cysteine-type endopeptidase inhibitor activity
protein binding
microtubule binding
Ran GTPase binding
enzyme binding
identical protein binding
metal ion binding
chaperone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000301633.8 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000350051.7 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000374948.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000586192.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000587746.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000589892.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590449.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000590925.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000591800.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592115.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 ENST00000592734.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012270.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001012271.1 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq
Affymetrix Human Clariom D Assay TC1700009043.hg.1 NM_001168.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline