This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05015260_g1
See other RPS6KB1 GE Assays ›
Gene Symbol:
RPS6KB1
Gene Name:
ribosomal protein S6 kinase B1
Gene Aliases:
PS6K, S6K, S6K-beta-1, S6K1, STK14A, p70 S6KA, p70(S6K)-alpha, p70-S6K, p70-alpha
Chromosome Location:
Chr.17: 59893046 - 59950426 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05015260_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS6KB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272042.1 NP_001258971.1 7 - 8 852 75
NM_001272043.1 NP_001258972.1 8 - 9 921 75
NM_001272044.1 NP_001258973.1 9 - 10 1050 75
NM_001272060.1 NP_001258989.1 8 - 9 921 75
NM_003161.3 NP_003152.1 8 - 9 921 75
XM_011525101.2 XP_011523403.1 8 - 9 916 75
XM_011525102.2 XP_011523404.1 8 - 9 916 75
XM_011525103.2 XP_011523405.1 10 - 11 1045 75
XM_017024929.1 XP_016880418.1 9 - 10 979 75
XM_017024930.1 XP_016880419.1 9 - 10 955 75
XM_017024931.1 XP_016880420.1 10 - 11 1169 75
XM_017024932.1 XP_016880421.1 9 - 10 1094 75
XM_017024933.1 XP_016880422.1 9 - 10 1089 75
GenBank mRNA AK293247.1 - 8 - 9 869 75
AK297147.1 - 9 - 10 857 75
AK300202.1 - 7 - 8 768 75
AK312875.1 - 8 - 9 827 75
BC036033.1 - 9 - 10 1153 75
BC053365.1 - 8 - 9 823 6142009 75
M60724.1 - 8 - 9 809 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPS6KB1

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
behavioral fear response
skeletal muscle contraction
protein phosphorylation
apoptotic process
signal transduction
germ cell development
aging
response to nutrient
long-term memory
response to heat
response to wounding
response to mechanical stimulus
response to toxic substance
response to glucose
skeletal muscle atrophy
response to electrical stimulus involved in regulation of muscle adaptation
positive regulation of smooth muscle cell migration
cell migration
TOR signaling
response to lipopolysaccharide
response to insulin
cellular response to hormone stimulus
response to testosterone
response to glucagon
response to tumor necrosis factor
response to drug
negative regulation of apoptotic process
response to leucine
protein kinase B signaling
response to ethanol
positive regulation of translation
positive regulation of mitotic cell cycle
positive regulation of translational initiation
regulation of glucose import
negative regulation of insulin receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of skeletal muscle tissue growth
positive regulation of smooth muscle cell proliferation
response to glucocorticoid
cellular response to growth factor stimulus
cellular response to organic cyclic compound
negative regulation of extrinsic apoptotic signaling pathway
protein kinase activity
ribosomal protein S6 kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
kinase activity
PDZ domain binding
peptide binding
identical protein binding
protein phosphatase 2A binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000441215.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 ENST00000578210.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq
Affymetrix Human Clariom D Assay TC1700007238.hg.1 NR_073509.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline