This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05016130_g1
See other EEF1A1 GE Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05016130_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001402.5 NP_001393.1 3 - 4 387 105
XM_011535514.2 XP_011533816.1 3 - 4 1022 105
GenBank mRNA AF174496.1 - 2 - 3 217 105
AF328730.1 - 1 95 105
AF397403.1 - 1 - 2 132 105
AJ420488.1 - 2 - 3 346 105
AK026650.1 - 1 387 105
AK055058.1 - 3 - 4 387 105
AK058051.1 - 3 - 4 387 105
AK058082.1 - 3 - 4 387 105
AK058163.1 - 2 - 3 205 105
AK098510.1 - 3 - 4 387 105
AK098518.1 - 3 - 4 247 105
AK098623.1 - 3 - 4 387 105
AK127384.1 - 3 - 4 387 105
AK222515.1 - 1 387 105
AK222523.1 - 1 387 105
AK222551.1 - 1 387 105
AK222982.1 - 1 387 105
AK223030.1 - 1 384 105
AK223046.1 - 1 390 105
AK289374.1 - 3 - 4 387 105
AK289442.1 - 3 - 4 325 105
AK290691.1 - 3 - 4 387 105
AK292639.1 - 3 - 4 703 105
AK297878.1 - 3 - 4 386 105
AK300922.1 - 3 - 4 248 105
AK304214.1 - 3 - 4 388 105
AK315827.1 - 3 - 4 378 105
AY043301.1 - 2 - 3 734 105
BC008587.1 - 1 373 3447654 105
BC009733.1 - 1 337 3877516 105
BC009875.2 - 1 377 3948601 105
BC010735.1 - 1 366 3898364 105
BC012891.1 - 1 365 3858483 105
BC014224.2 - 1 376 4335756 105
BC018150.1 - 1 373 3851939 105
BC018641.2 - 1 373 3660140 105
BC018850.2 - 1 376 105
BC019050.2 - 1 339 105
BC021686.1 - 1 392 3930863 105
BC028674.1 - 1 391 4107346 105
BC029337.1 - 1 361 105
BC029343.1 - 1 359 105
BC029997.1 - 1 373 105
BC035877.1 - 1 388 105
BC038339.1 - 1 373 3461588 105
BC057391.1 - 1 342 4339606 105
BC066893.1 - 1 387 4818863 105
BC071619.1 - 1 366 4448926 105
BC071727.1 - 1 342 4514479 105
BC071741.1 - 1 367 3885539 105
BC071841.1 - 1 362 4587981 105
BC072385.1 - 1 374 5585092 105
BC082268.1 - 1 366 6729595 105
BC111051.1 - 1 377 5585079 105
DQ867534.1 - 1 73 105
EF219140.1 - 3 - 4 547 105
EF362804.1 - 3 - 4 617 105
X03558.1 - 3 - 4 377 105
X16869.1 - 2 - 3 339 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF328730.1, AK289442.1, AK304214.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1A1

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0