This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05017131_s1
See other IGF2BP1 GE Assays ›
Gene Symbol:
IGF2BP1
Gene Name:
insulin like growth factor 2 mRNA binding protein 1
Gene Aliases:
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1
Chromosome Location:
Chr.17: 48997412 - 49056145 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05017131_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGF2BP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160423.1 NP_001153895.1 13 4846 116
NM_006546.3 NP_006537.3 15 5263 116
XM_005256955.3 XP_005257012.2 14 4986 116
XM_011524201.2 XP_011522503.1 15 5211 116
XM_017024022.1 XP_016879511.1 15 5208 116
GenBank mRNA AK095573.1 - 1 825 116

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGF2BP1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

gene expression
regulation of mRNA stability involved in response to stress
negative regulation of translation
pallium cell proliferation in forebrain
regulation of cytokine biosynthetic process
mRNA transport
CRD-mediated mRNA stabilization
neuronal stem cell population maintenance
nucleotide binding
mRNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000578722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000578722.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline