This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05018265_m1
See other DR1 GE Assays ›
Gene Symbol:
DR1
Gene Name:
down-regulator of transcription 1
Gene Aliases:
NC2, NC2-BETA, NC2B
Chromosome Location:
Chr.1: 93345921 - 93362592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05018265_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL137673.1 - 1 - 2 794 128

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000481583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000481583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 ENST00000481583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370267.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000370272.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000481583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000481583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 ENST00000481583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq
Affymetrix Human Clariom D Assay TC0100009061.hg.1 NM_001938.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0