This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05018343_mH
See other ATP5F1 GE Assays ›
Gene Symbol:
ATP5F1
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
Gene Aliases:
PIG47
Chromosome Location:
Chr.1: 111449121 - 111461918 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05018343_mH
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5F1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001688.4 NP_001679.2 4 - 5 813 130
GenBank mRNA AK223018.1 - 4 - 5 438 130
AK291077.1 - 4 - 5 470 130
AK308266.1 - 5 - 6 850 131
AK313541.1 - 4 - 5 440 130
AY871272.1 - 4 - 5 447 130
BC005366.1 - 4 - 5 458 3839745 130
BC005960.1 - 4 - 5 450 4049491 130
BC016350.1 - 4 - 5 489 4074170 130
X60221.1 - 4 - 5 424 130

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5F1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
ATP synthesis coupled proton transport
substantia nigra development
mitochondrial ATP synthesis coupled proton transport
protein binding
ATPase activity
transmembrane transporter activity
proton-transporting ATP synthase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369721.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000369722.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000464154.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000468818.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000483994.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 ENST00000493119.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq
Affymetrix Human Clariom D Assay TC0100009414.hg.1 NM_001688.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0