This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05018469_m1
See other GPSM2 GE Assays ›
Gene Symbol:
GPSM2
Gene Name:
G-protein signaling modulator 2
Gene Aliases:
CMCS, DFNB82, LGN, PINS
Chromosome Location:
Chr.1: 108875350 - 108934335 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05018469_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPSM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321038.1 NP_001307967.1 2 - 3 320 87
NM_001321039.1 NP_001307968.1 2 - 3 552 87
NM_013296.4 NP_037428.3 2 - 3 552 87
XM_011541301.2 XP_011539603.1 3 - 4 829 87
XM_011541302.2 XP_011539604.1 3 - 4 1491 87
XM_011541303.2 XP_011539605.1 2 - 3 1247 87
XM_017001097.1 XP_016856586.1 3 - 4 1397 87
XM_017001098.1 XP_016856587.1 3 - 4 1165 87
GenBank mRNA AB445462.1 - 2 - 3 552 87
AK295563.1 - 2 - 3 139 87
AK314249.1 - 2 - 3 548 87
BC027732.2 - 2 - 3 328 5020191 87
DC341689.1 - 3 - 4 366 87
DC365139.1 - 2 - 3 320 87
KX085485.1 - 2 - 3 552 87
U54999.1 - 1 - 2 208 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB445462.1, AK295563.1, AK314249.1, BC027732.2, NM_001321038.1, NM_001321039.1, NM_013296.4, U54999.1, XM_011541301.2, XM_011541302.2, XM_011541303.2, XM_017001097.1, XM_017001098.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPSM2

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
mitotic spindle organization
G-protein coupled receptor signaling pathway
regulation of catalytic activity
maintenance of centrosome location
lung epithelial cell differentiation
nucleotide binding
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
protein binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000264126.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000406462.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435475.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000435987.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000441735.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 ENST00000446797.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq
Affymetrix Human Clariom D Assay TC0100009333.hg.1 NM_013296.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0