This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05022281_mH
See other GSTM1 GE Assays ›
Gene Symbol:
GSTM1
Gene Name:
glutathione S-transferase mu 1
Gene Aliases:
GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, MU, MU-1
Chromosome Location:
Chr.1: 109687796 - 109693745 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05022281_mH
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA KJ534856.1 - 5 - 6 360 64
KJ534998.1 - 4 - 5 278 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000309851.9 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000349334.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369819.2 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000369823.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000476065.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 ENST00000490171.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_000561.3 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq
Affymetrix Human Clariom D Assay TC0100018263.hg.1 NM_146421.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0