This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05022320_m1
See other ATPAF1 GE Assays ›
Gene Symbol:
ATPAF1
Gene Name:
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases:
ATP11, ATP11p
Chromosome Location:
Chr.1: 46635039 - 46668427 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05022320_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATPAF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256418.1 NP_001243347.1 3 - 4 434 63
GenBank mRNA AK301874.1 - 3 - 4 411 63
AK310338.1 - 3 - 4 404 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK301874.1, NM_001256418.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial proton-transporting ATP synthase complex assembly
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mz2gr:80/100.66.78.42:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0