This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05024048_g1
See other RBM17 GE Assays ›
Gene Symbol:
RBM17
Gene Name:
RNA binding motif protein 17
Gene Aliases:
SPF45
Chromosome Location:
Chr.10: 6088986 - 6117459 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05024048_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145547.1 NP_001139019.1 10 - 11 1670 102
NM_032905.4 NP_116294.1 10 - 11 1252 102
GenBank mRNA AF542550.1 - 10 - 11 1194 102
AK314952.1 - 10 - 11 1176 102
BC007871.2 - 10 - 11 1164 4303675 102
BC009064.1 - 9 - 10 931 102
BC039322.1 - 10 - 11 1673 5267008 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM17

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

alternative mRNA splicing, via spliceosome
nucleotide binding
RNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0