This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05024066_m1
See other ZEB1 GE Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05024066_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZEB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323650.1 NP_001310579.1 1 - 2 126 78
NM_001323653.1 NP_001310582.1 1 - 2 126 78
NM_001323655.1 NP_001310584.1 1 - 2 126 78
XM_006717498.2 XP_006717561.1 1 - 2 100 78
XM_017016597.1 XP_016872086.1 1 - 2 85 78
GenBank mRNA AL831979.1 - 1 - 2 82 78
BX954549.1 - 1 - 2 84 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001323650.1, NM_001323653.1, NM_001323655.1, XM_006717498.2, XM_017016597.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline