This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05024379_s1
See other ITCH GE Assays ›
Gene Symbol:
ITCH
Gene Name:
itchy E3 ubiquitin protein ligase
Gene Aliases:
ADMFD, AIF4, AIP4, NAPP1
Chromosome Location:
Chr.20: 34363235 - 34511773 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05024379_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ITCH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257137.2 NP_001244066.1 26 4816 111
NM_001257138.2 NP_001244067.1 24 4551 111
NM_001324197.1 NP_001311126.1 26 4837 111
NM_001324198.1 NP_001311127.1 25 4714 111
NM_031483.6 NP_113671.3 25 4693 111
XM_005260570.1 XP_005260627.1 24 4609 111
XM_011529079.1 XP_011527381.1 24 4629 111
XM_017028089.1 XP_016883578.1 25 4649 111
XM_017028090.1 XP_016883579.1 26 4769 111
XM_017028091.1 XP_016883580.1 24 4564 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ308022.1 - 1 Chr. 20 - 34509543 - 34510010 [-] on Build GRCh38 111

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ITCH

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
positive regulation of T cell anergy
ubiquitin-dependent protein catabolic process
apoptotic process
inflammatory response
Notch signaling pathway
protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
negative regulation of type I interferon production
protein K29-linked ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of apoptotic process
innate immune response
positive regulation of protein catabolic process
negative regulation of JNK cascade
negative regulation of alpha-beta T cell proliferation
viral entry into host cell
negative regulation of defense response to virus
defense response to virus
nucleotide-binding oligomerization domain containing signaling pathway
protein K63-linked ubiquitination
protein K48-linked ubiquitination
regulation of protein deubiquitination
ubiquitin-protein transferase activity
protein binding
ligase activity
ubiquitin-like protein transferase activity
ribonucleoprotein complex binding
CXCR chemokine receptor binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0