This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05025071_g1
See other CHAF1B GE Assays ›
Gene Symbol:
CHAF1B
Gene Name:
chromatin assembly factor 1 subunit B
Gene Aliases:
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
Chromosome Location:
Chr.21: 36380145 - 36416827 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05025071_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHAF1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017028478.1 XP_016883967.1 2 - 3 1232 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017028478.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
DNA replication
DNA repair
DNA replication-dependent nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
protein complex assembly
cell cycle
chromatin assembly
mitophagy in response to mitochondrial depolarization
xenophagy
chromatin binding
histone binding
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline