This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05025083_m1
See other IGSF5 GE Assays ›
Gene Symbol:
IGSF5
Gene Name:
immunoglobulin superfamily member 5
Gene Aliases:
GSF5, JAM4
Chromosome Location:
Chr.21: 39725838 - 39802096 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05025083_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011529472.1 XP_011527774.1 1 - 2 251 124
XM_011529473.1 XP_011527775.1 1 - 2 251 124
XM_017028279.1 XP_016883768.1 1 - 2 251 124
XM_017028280.1 XP_016883769.1 1 - 2 251 124

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011529472.1, XM_011529473.1, XM_017028279.1, XM_017028280.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin

Gene Ontology Categories:

Function(s)

single organismal cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007172.hg.1 ENST00000416555.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007172.hg.1 ENST00000416555.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0