This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05025899_m1
See other NDUFV3 GE Assays ›
Gene Symbol:
NDUFV3
Gene Name:
NADH:ubiquinone oxidoreductase subunit V3
Gene Aliases:
CI-10k, CI-9KD
Chromosome Location:
Chr.21: 42893268 - 42911396 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05025899_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFV3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017028359.1 XP_016883848.1 2 - 3 227 66
GenBank mRNA AY173949.1 - 1 - 2 127 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0