This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05026755_m1
See other DHRS9 GE Assays ›
Gene Symbol:
DHRS9
Gene Name:
dehydrogenase/reductase 9
Gene Aliases:
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4
Chromosome Location:
Chr.2: 169067035 - 169096167 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05026755_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF240697.1 - 1 - 2 63 88
DB010228.1 - 1 - 2 156 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF240697.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline