This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05029807_s1
See other HDAC2 GE Assays ›
Gene Symbol:
HDAC2
Gene Name:
histone deacetylase 2
Gene Aliases:
HD2, RPD3, YAF1
Chromosome Location:
Chr.6: 113936156 - 113971195 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05029807_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001527.3 NP_001518.3 14 2811 71
NR_033441.1 15 2839 71
NR_073443.1 14 2717 71
GenBank mRNA CN304190.1 - 1 266 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HDAC2

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
cardiac muscle hypertrophy
chromatin remodeling
maintenance of chromatin silencing
transcription, DNA-templated
blood coagulation
positive regulation of cell proliferation
epidermal cell differentiation
positive regulation of epithelial to mesenchymal transition
positive regulation of receptor biosynthetic process
negative regulation of neuron projection development
dendrite development
histone deacetylation
response to caffeine
response to lipopolysaccharide
positive regulation of interleukin-1 production
positive regulation of tumor necrosis factor production
circadian regulation of gene expression
positive regulation of collagen biosynthetic process
cellular response to heat
response to nicotine
response to cocaine
odontogenesis of dentin-containing tooth
response to drug
positive regulation of tyrosine phosphorylation of Stat3 protein
embryonic digit morphogenesis
ATP-dependent chromatin remodeling
negative regulation of apoptotic process
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of MHC class II biosynthetic process
positive regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
behavioral response to ethanol
positive regulation of oligodendrocyte differentiation
response to hyperoxia
hair follicle placode formation
negative regulation of dendritic spine development
eyelid development in camera-type eye
fungiform papilla formation
cellular response to hydrogen peroxide
histone H3 deacetylation
histone H4 deacetylation
cellular response to retinoic acid
cellular response to transforming growth factor beta stimulus
regulation of signal transduction by p53 class mediator
cellular response to dopamine
negative regulation of peptidyl-lysine acetylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
core promoter binding
RNA polymerase II repressing transcription factor binding
chromatin binding
histone deacetylase activity
protein binding
transcription factor binding
deacetylase activity
enzyme binding
heat shock protein binding
nucleosomal DNA binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
sequence-specific DNA binding
poly(A) RNA binding
NF-kappaB binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600006522.hg.1 ENST00000561511.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006522.hg.1 ENST00000561511.1 Ensembl
Affymetrix Human Clariom D Assay TC1600006522.hg.1 ENST00000561511.1 Ensembl
Affymetrix Human Clariom D Assay TC1600006522.hg.1 ENST00000561511.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0