This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05030024_s1
See other TAP2 GE Assays ›
Gene Symbol:
TAP2
Gene Name:
transporter 2, ATP binding cassette subfamily B member
Gene Aliases:
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
Chromosome Location:
Chr.6: 32821833 - 32838823 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05030024_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000544.3 NP_000535.3 12 4342 93
NM_001290043.1 NP_001276972.1 12 4395 93
GenBank mRNA AB208953.1 - 13 4122 93
AL833658.1 - 1 1599 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAP2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
intracellular transport of viral protein in host cell
antigen processing and presentation of endogenous peptide antigen via MHC class I
cytosol to ER transport
peptide antigen transport
transmembrane transport
transporter activity
protein binding
ATP binding
peptide transporter activity
peptide antigen-transporting ATPase activity
MHC class Ib protein binding
TAP1 binding
tapasin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline