This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05030278_s1
See other MYB GE Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05030278_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130173.1 NP_001123645.1 10 1713 100
NM_001161656.1 NP_001155128.1 10 1704 100
NM_001161658.1 NP_001155130.1 10 1665 100
GenBank mRNA AJ606319.1 - 10 1713 100
AJ616799.2 - 3 749 100
AY787455.1 - 11 1645 100
AY787460.1 - 11 1627 100
AY787461.1 - 10 1505 100
AY787462.1 - 10 1654 100
AY787463.1 - 10 1420 100
AY787465.1 - 10 1748 100
AY787467.1 - 10 1514 100
AY787468.1 - 10 1466 100
AY787469.1 - 10 1466 100
AY787473.1 - 10 1663 100
AY787474.1 - 10 1429 100
X17469.1 - 1 323 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0