This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05032316_s1
See other BATF3 GE Assays ›
Gene Symbol:
BATF3
Gene Name:
basic leucine zipper ATF-like transcription factor 3
Gene Aliases:
JDP1, JUNDM1, SNFT
Chromosome Location:
Chr.1: 212686417 - 212699985 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05032316_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BATF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018664.2 NP_061134.1 3 797 119
GenBank mRNA AF255346.1 - 3 797 119
AK093671.1 - 2 2174 119
AK094854.1 - 2 1973 119
BG184570.1 - 2 424 119

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BATF3

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
response to virus
myeloid dendritic cell differentiation
dendritic cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000243440.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000471382.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 ENST00000478275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq
Affymetrix Human Clariom D Assay TC0100017258.hg.1 NM_018664.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0