This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05032864_g1
See other PRSS3 GE Assays ›
Gene Symbol:
PRSS3
Gene Name:
protease, serine 3
Gene Aliases:
MTG, PRSS4, T9, TRY3, TRY4
Chromosome Location:
Chr.9: 33750466 - 33799231 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05032864_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197097.2 NP_001184026.2 1 174 115
NM_007343.3 NP_031369.2 1 174 115
XM_011517965.1 XP_011516267.1 1 118 115
GenBank mRNA AB298285.1 - 1 174 115

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001197097.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
cobalamin metabolic process
zymogen activation
endothelial cell migration
serine-type endopeptidase activity
calcium ion binding
protein binding
serine-type peptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000477653.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000477653.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000477653.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000342836.8 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000361005.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000379405.3 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000429677.7 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000457896.5 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000468152.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000477653.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000477653.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000477653.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 ENST00000495682.1 Ensembl
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197097.2 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_001197098.1 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_002771.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq
Affymetrix Human Clariom D Assay TC0900007006.hg.1 NM_007343.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline