This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05035099_s1
See other GJB6 GE Assays ›
Gene Symbol:
GJB6
Gene Name:
gap junction protein beta 6
Gene Aliases:
CX30, DFNA3, DFNA3B, DFNB1B, ECTD2, ED2, EDH, HED, HED2
Chromosome Location:
Chr.13: 20221962 - 20232395 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05035099_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110219.2 NP_001103689.1 4 492 100
NM_001110220.2 NP_001103690.1 3 382 100
NM_001110221.2 NP_001103691.1 2 258 100
NM_006783.4 NP_006774.2 2 424 100
XM_017020357.1 XP_016875846.1 3 697 100
XM_017020358.1 XP_016875847.1 4 387 100
XM_017020359.1 XP_016875848.1 4 417 100
XM_017020360.1 XP_016875849.1 3 184 100
GenBank mRNA AK075247.1 - 4 324 100
AK289592.1 - 2 172 100
AK312352.1 - 2 130 101
AY789474.1 - 2 96 100
AY789475.1 - 3 220 100
AY789476.1 - 4 336 100
BC038934.1 - 2 119 5196769 100
BP227986.1 - 2 180 100
DC295753.1 - 3 300 100
DC337465.1 - 2 424 100
DN998505.1 - 4 494 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK075247.1, AK289592.1, AY789474.1, AY789475.1, AY789476.1, BC038934.1, NM_001110219.2, NM_001110220.2, NM_001110221.2, NM_006783.4, XM_017020357.1, XM_017020358.1, XM_017020359.1, XM_017020360.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GJB6

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s)

apoptotic process
cell communication
aging
sensory perception of sound
negative regulation of cell proliferation
response to lipopolysaccharide
ear morphogenesis
inner ear development
response to electrical stimulus
cellular response to glucose stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000241124.10 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000356192.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400065.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 ENST00000400066.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110219.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110220.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_001110221.2 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq
Affymetrix Human Clariom D Assay TC1300008250.hg.1 NM_006783.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline