This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05035101_s1
See other EEF1AKMT1 GE Assays ›
Gene Symbol:
EEF1AKMT1
Gene Name:
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1
Gene Aliases:
ESP13, N6AMT2
Chromosome Location:
Chr.13: 20728931 - 20773958 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05035101_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1AKMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA HY004043.1 - 1 183 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline