This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05036194_s1
See other DISP2 GE Assays ›
Gene Symbol:
DISP2 LINC00594
Gene Name:
dispatched RND transporter family member 2
long intergenic non-protein coding RNA 594
Gene Aliases:
DISPB, HsT16908, LINC00594
C15orf36
Chromosome Location:
Chr.15: 40354359 - 40378755 on Build GRCh38
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05036194_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DISP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011522125.2 XP_011520427.1 9 8192 100
XM_017022687.1 XP_016878176.1 9 8192 100
Gene Symbol: LINC00594
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AI278850.1 - 2 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

smoothened signaling pathway
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500010718.hg.1 ENST00000561261.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010718.hg.1 ENST00000561261.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010718.hg.1 ENST00000561261.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010718.hg.1 ENST00000561261.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010718.hg.1 ENST00000561261.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010718.hg.1 ENST00000561261.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0