This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05037736_s1
See other SUPT6H GE Assays ›
Gene Symbol:
SUPT6H
Gene Name:
SPT6 homolog, histone chaperone
Gene Aliases:
SPT6, SPT6H, emb-5
Chromosome Location:
Chr.17: 28662183 - 28702684 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05037736_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUPT6H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320755.1 NP_001307684.1 23 3029 79
NM_003170.4 NP_003161.2 22 2927 79
XM_017024978.1 XP_016880467.1 24 3088 79
XM_017024979.1 XP_016880468.1 22 3165 79
XM_017024980.1 XP_016880469.1 21 2859 79
GenBank mRNA AL833607.1 - 23 3030 79
BC003696.1 - 7 802 79
BC017105.1 - 2 70 79
BC136522.1 - 22 2839 9020512 79
BC136524.1 - 22 2839 9020514 79
BC150268.1 - 22 2841 9007230 79
D79984.1 - 22 2841 79
U38658.1 - 2 123 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SUPT6H

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
regulation of mRNA export from nucleus
viral process
positive regulation of transcription elongation from RNA polymerase II promoter
regulation of isotype switching
regulation of mRNA processing
mRNA transport
regulation of muscle cell differentiation
negative regulation of histone H3-K27 methylation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
histone binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0