This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05041626_s1
See other YIF1B GE Assays ›
Gene Symbol:
YIF1B
Gene Name:
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases:
FinGER8
Chromosome Location:
Chr.19: 38303560 - 38315976 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05041626_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YIF1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039671.2 NP_001034760.1 8 1323 71
NM_001039672.2 NP_001034761.1 8 1341 71
NM_001039673.2 NP_001034762.1 8 1332 71
NM_001145461.1 NP_001138933.1 8 1317 71
NM_001145462.1 NP_001138934.1 8 1293 71
NM_001145463.1 NP_001138935.1 9 1409 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000301246.9 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000588605.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000589986.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 ENST00000591852.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Clariom D Assay TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Clariom D Assay TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Clariom D Assay TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Clariom D Assay TC1900011726.hg.1 NM_033520.2 Refseq
Affymetrix Human Clariom D Assay TC1900011726.hg.1 NM_033520.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0