This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05042351_s1
See other CADM1 GE Assays ›
Gene Symbol:
CADM1
Gene Name:
cell adhesion molecule 1
Gene Aliases:
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
Chromosome Location:
Chr.11: 115173625 - 115504523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05042351_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CADM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098517.1 NP_001091987.1 9 2700 106
NM_001301043.1 NP_001287972.1 12 2871 106
NM_001301044.1 NP_001287973.1 11 2817 106
NM_001301045.1 NP_001287974.1 11 2787 106
NM_014333.3 NP_055148.3 10 2784 106
XM_005271494.2 XP_005271551.1 10 2626 106
XM_017017457.1 XP_016872946.1 12 2830 106
XM_017017458.1 XP_016872947.1 10 2550 106
XM_017017459.1 XP_016872948.1 13 4824 106
XM_017017460.1 XP_016872949.1 11 4737 106
XM_017017461.1 XP_016872950.1 10 2667 106
GenBank mRNA AF132811.1 - 10 2661 106
AK172730.1 - 1 183 106
BM309987.1 - 1 318 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CADM1

Gene Ontology Categories:

Function(s) Process(es)

T cell mediated cytotoxicity
apoptotic process
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
multicellular organism development
spermatogenesis
cell recognition
cell differentiation
adherens junction organization
susceptibility to natural killer cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
activated T cell proliferation
detection of stimulus
receptor activity
receptor binding
protein binding
PDZ domain binding
protein homodimerization activity
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lwb2x:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0