This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05042946_s1
See other MCEE GE Assays ›
Gene Symbol:
MCEE
Gene Name:
methylmalonyl-CoA epimerase
Gene Aliases:
GLOD2
Chromosome Location:
Chr.2: 71109676 - 71130288 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05042946_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MCEE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032601.3 NP_115990.3 3 552 146
XM_005264613.2 XP_005264670.1 3 414 146
GenBank mRNA AB209469.1 - 1 5603 146
AI033764.1 - 1 146
BC020825.1 - 3 517 4734422 146

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MCEE

Panther Classification:

Molecular Function -

epimerase/racemase isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

short-chain fatty acid catabolic process
L-methylmalonyl-CoA metabolic process
methylmalonyl-CoA epimerase activity
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000244217.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000413592.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000462609.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000486135.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 ENST00000494660.6 Ensembl
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq
Affymetrix Human Clariom D Assay TC0200013019.hg.1 NM_032601.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline