This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05043311_g1
See other IGSF3 GE Assays ›
Gene Symbol:
IGSF3
Gene Name:
immunoglobulin superfamily member 3
Gene Aliases:
EWI-3, LCDD, V8
Chromosome Location:
Chr.1: 116574398 - 116667755 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05043311_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007237.2 NP_001007238.1 6 2164 93
NM_001542.3 NP_001533.2 6 2086 93
XM_005270793.2 XP_005270850.1 7 2196 93
XM_005270794.4 XP_005270851.1 5 1434 93
XM_006710593.3 XP_006710656.1 6 1497 93
XM_011541315.1 XP_011539617.1 7 2248 93
GenBank mRNA AB007935.2 - 5 1496 93
AF031174.1 - 6 1561 93
AK097809.1 - 2 496 93
BC110651.1 - 6 2164 6166346 93
BC130279.1 - 5 1834 40146310 93
BC144132.1 - 5 1834 9052652 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF3

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

lacrimal gland development
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0