This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05043692_s1
See other ADPRH GE Assays ›
Gene Symbol:
ADPRH
Gene Name:
ADP-ribosylarginine hydrolase
Gene Aliases:
ARH1
Chromosome Location:
Chr.3: 119579433 - 119589954 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05043692_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADPRH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001125.3 NP_001116.1 1 221 108
NM_001291949.1 NP_001278878.1 1 221 108
NM_001291950.1 NP_001278879.1 1 221 108
XM_011512438.2 XP_011510740.1 1 221 108
GenBank mRNA AK291787.1 - 1 64 108
AK304560.1 - 1 126 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK291787.1, NM_001125.3, NM_001291949.1, NM_001291950.1, XM_011512438.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADPRH

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
intracellular protein transport
regulation of vesicle fusion
protein de-ADP-ribosylation
activation of GTPase activity
magnesium ion binding
ADP-ribosylarginine hydrolase activity
GTPase activator activity
protein binding
Rab GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline