This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05044248_g1
See other PPP1R2 GE Assays ›
Gene Symbol:
PPP1R2
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 2
Gene Aliases:
IPP-2, IPP2
Chromosome Location:
Chr.3: 195514422 - 195543401 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05044248_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291504.1 NP_001278433.1 1 165 103
NM_001291505.1 NP_001278434.1 1 165 103
NM_001316325.1 NP_001303254.1 1 165 103
NM_006241.7 NP_006232.1 1 165 103
NR_133014.1 1 165 103
XM_006713682.3 XP_006713745.1 1 112 103
GenBank mRNA BC066921.1 - 1 150 103
BI549862.1 - 1 83 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC066921.1, NM_001291504.1, NM_001291505.1, NM_001316325.1, NM_006241.7, XM_006713682.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R2

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase inhibitor phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

glycogen metabolic process
generation of precursor metabolites and energy
regulation of signal transduction
negative regulation of catalytic activity
regulation of phosphoprotein phosphatase activity
protein serine/threonine phosphatase inhibitor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700009366.hg.1 ENST00000478565.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000625807.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000625807.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 ENST00000625807.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0700009366.hg.1 ENST00000478565.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000413183.3 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000418939.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000438848.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000460437.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000462906.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000618156.4 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000625807.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000625807.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 ENST00000625807.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291504.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_001291505.1 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq
Affymetrix Human Clariom D Assay TC0300013639.hg.1 NM_006241.7 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0