This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05044585_s1
See other CYTL1 GE Assays ›
Gene Symbol:
CYTL1
Gene Name:
cytokine like 1
Gene Aliases:
C17, C4orf4
Chromosome Location:
Chr.4: 5014586 - 5019470 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05044585_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYTL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018659.2 NP_061129.1 4 625 94
XM_017008299.1 XP_016863788.1 5 740 94
GenBank mRNA AF193766.1 - 4 628 94
AF274956.1 - 4 485 94
AY359101.1 - 4 626 94
BC031391.1 - 4 601 5186334 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CYTL1

Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

chondrocyte differentiation
signal transduction
positive regulation of transcription from RNA polymerase II promoter
inner ear development
chondroitin sulfate proteoglycan biosynthetic process
positive regulation of sequence-specific DNA binding transcription factor activity
cartilage homeostasis
receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000506508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000506508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000506508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000509419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000509419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 ENST00000509419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000307746.8 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000506508.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000506508.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000506508.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000509419.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000509419.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 ENST00000509419.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Clariom D Assay TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Clariom D Assay TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Clariom D Assay TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Clariom D Assay TC0400009879.hg.1 NM_018659.2 Refseq
Affymetrix Human Clariom D Assay TC0400009879.hg.1 NM_018659.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline