This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05046073_s1
See other NRM GE Assays ›
Gene Symbol:
NRM
Gene Name:
nurim (nuclear envelope membrane protein)
Gene Aliases:
NRM29
Chromosome Location:
Chr.6: 30688047 - 30691420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05046073_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NRM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270707.1 NP_001257636.1 2 394 69
NM_001270708.1 NP_001257637.1 2 394 69
NM_001270709.1 NP_001257638.1 2 394 69
NM_007243.2 NP_009174.1 3 591 69
NR_073065.1 2 394 69
NR_073072.1 2 237 69
XM_017010224.1 XP_016865713.1 2 593 69
XM_017010225.1 XP_016865714.1 3 507 69
GenBank mRNA AF143676.1 - 2 237 69
AK075509.1 - 3 593 69
AK315190.1 - 2 249 69
AY358411.1 - 2 258 69
BC021260.1 - 2 229 69
BC039865.1 - 2 257 6043663 69
BE548635.1 - 2 236 69
BM847576.1 - 2 268 69
BM917173.1 - 2 251 69
BU159435.1 - 2 203 69
JQ798186.1 - 2 257 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017010224.1, XM_017010225.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NRM

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0