This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05046781_s1
See other GPR85 GE Assays ›
Gene Symbol:
GPR85
Gene Name:
G protein-coupled receptor 85
Gene Aliases:
SREB, SREB2
Chromosome Location:
Chr.7: 113080413 - 113087778 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05046781_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146265.1 NP_001139737.1 3 925 67
NM_001146266.1 NP_001139738.1 2 815 67
NM_001146267.1 NP_001139739.1 3 752 67
NM_018970.6 NP_061843.3 3 922 67
XM_005250451.2 XP_005250508.1 2 872 67
XM_017012352.1 XP_016867841.1 3 987 67
GenBank mRNA AB040800.1 - 1 298 67
AF250237.2 - 3 910 67
AK312587.1 - 2 690 67
AL161959.1 - 3 755 67
BC030577.1 - 3 876 4823671 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR85

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000297146.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000424100.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000438062.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449591.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000449735.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000487573.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 ENST00000610164.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146265.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146266.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_001146267.1 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq
Affymetrix Human Clariom D Assay TC0700012299.hg.1 NM_018970.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline