This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05048179_s1
See other GNE GE Assays ›
Gene Symbol:
GNE
Gene Name:
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases:
DMRV, GLCNE, IBM2, NM, Uae1
Chromosome Location:
Chr.9: 36214441 - 36277056 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05048179_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128227.2 NP_001121699.1 3 470 121
NM_001190383.1 NP_001177312.1 3 486 121
NM_001190384.1 NP_001177313.1 2 280 121
NM_001190388.1 NP_001177317.1 2 264 121
NM_005476.5 NP_005467.1 3 486 121
XM_005251334.4 XP_005251391.1 3 470 121
XM_017014167.1 XP_016869656.1 3 331 121
XM_017014168.1 XP_016869657.1 3 463 121
GenBank mRNA AF051852.1 - 2 315 121
AF155663.1 - 2 265 121
AJ238764.1 - 2 306 121
AK295562.1 - 2 226 121
AK296687.1 - 2 181 121
AK312539.1 - 3 400 121
AL701451.1 - 3 392 121
AM697708.1 - 3 358 121
AM697709.1 - 2 142 121
AY531126.1 - 2 306 121
AY531127.1 - 2 306 121
AY531128.1 - 2 306 121
BC121179.2 - 3 394 40122902 121
BP201585.1 - 3 486 121
DC416534.1 - 3 458 121
DC418354.1 - 3 471 121
EU093084.1 - 2 316 121

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GNE

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

N-acetylglucosamine biosynthetic process
UDP-N-acetylglucosamine metabolic process
N-acetylneuraminate metabolic process
cell adhesion
carbohydrate phosphorylation
hydrolase activity, hydrolyzing O-glycosyl compounds
protein binding
ATP binding
UDP-N-acetylglucosamine 2-epimerase activity
N-acylmannosamine kinase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline