This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05049173_g1
See other PPP1R12B GE Assays ›
Gene Symbol:
PPP1R12B
Gene Name:
protein phosphatase 1 regulatory subunit 12B
Gene Aliases:
MYPT2, PP1bp55
Chromosome Location:
Chr.1: 202348702 - 202588569 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05049173_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R12B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002481.3 NP_002472.2 16 2427 79
NM_032103.2 NP_115286.1 4 782 79
NM_032104.2 NP_115287.1 4 782 79
XM_017001342.1 XP_016856831.1 17 2610 79
XM_017001343.1 XP_016856832.1 17 2610 79
XM_017001344.1 XP_016856833.1 17 2610 79
XM_017001345.1 XP_016856834.1 17 2610 79
XM_017001346.1 XP_016856835.1 16 2427 79
XM_017001347.1 XP_016856836.1 16 2427 79
XM_017001348.1 XP_016856837.1 16 2427 79
XM_017001349.1 XP_016856838.1 17 2610 79
XM_017001350.1 XP_016856839.1 17 2484 79
XM_017001352.1 XP_016856841.1 7 877 79
XM_017001353.1 XP_016856842.1 7 867 79
XM_017001354.1 XP_016856843.1 4 2261 79
GenBank mRNA AB003062.1 - 16 2440 79
AB050641.1 - 4 782 79
AB050642.1 - 4 782 79
AF324888.1 - 16 2421 79
AK307591.1 - 16 2427 79
BC144699.1 - 16 2419 9053233 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB050641.1, AB050642.1, NM_032103.2, NM_032104.2, XM_017001352.1, XM_017001353.1, XM_017001354.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R12B

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
regulation of muscle contraction
signal transduction
positive regulation of catalytic activity
protein binding
enzyme activator activity
phosphatase regulator activity
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline