This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05049447_s1
See other ORAI3 GE Assays ›
Gene Symbol:
ORAI3
Gene Name:
ORAI calcium release-activated calcium modulator 3
Gene Aliases:
TMEM142C
Chromosome Location:
Chr.16: 30949084 - 30954938 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05049447_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ORAI3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152288.2 NP_689501.1 1 318 76
GenBank mRNA AK298276.1 - 1 280 76
BC006126.1 - 1 261 3603998 76
BC015555.1 - 1 307 3878011 76
BC022786.1 - 1 261 3605904 76
BI561433.1 - 1 339 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ORAI3

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
calcium ion transmembrane transport
protein binding
store-operated calcium channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000562699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000562699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000562699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000318663.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000562699.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000562699.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000562699.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000563161.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 ENST00000566237.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq
Affymetrix Human Clariom D Assay TC1600011386.hg.1 NM_152288.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0